EMAIL THIS PAGE TO A FRIEND
74415 Fluka

Ammonium carbonate

for HPLC, ≥30% NH3 basis (T)

Synonym: Hartshorn salt

  • CAS Number 506-87-6

  • Linear Formula (NH4)2CO3

  • Molecular Weight 96.09

  •  Beilstein Registry Number 4253487

  •  EC Number 208-058-0

  •  MDL number MFCD00010890

  •  PubChem Substance ID 24886691

  •  eCl@ss 38020208

  • Popular Documents:  FTIR (PDF)  

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Buffers & Reagents, Chromatography Reagents & Solvents, HPLC,
grade   for HPLC
vapor density   2.7 (vs air)
assay   ≥30% NH3 basis (T)
solubility   H2O: soluble0.1 g/mL at 25 °C, clear, colorless
λ   10 % in H2O
UV absorption   λ: 240 nm Amax: 0.1
  λ: 250 nm Amax: 0.04
  λ: 260 nm Amax: 0.02
  λ: 280 nm Amax: 0.01
  λ: 500 nm Amax: 0.01
suitability   complies for cyclovoltagram
  corresponds for filter test
  passes test for RP gradient test

Description

Legal Information

FLUKA is a registered trademark of Honeywell Specialty Chemicals Seelze GmbH

Application

Ammonium carbonate was used in experiment to distinguish 26 different phenolic components and in Capillary electrophoresis-mass spectrometry (CE-MS/MS) as rapid screening technique.

General description

Ammonium carbonate is a hard white translucent mass. It is suitable as a leavening agent, pH-control agent, dough strengthener and texturizer. It can be prepared by ammonium sulphate and calcium carbonate by sublimation.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
BP1925000
Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?