74691 Fluka

Octadecane

analytical standard

DOWNLOAD MSDS (PDF)
  • CAS Number 593-45-3

  • Linear Formula CH3(CH2)16CH3

  • Molecular Weight 254.49

  •  Beilstein Registry Number 1740138

  •  EC Number 209-790-3

  •  MDL number MFCD00009007

  •  PubChem Substance ID 24886704

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chemical Class, Hydrocarbons,
grade   analytical standard
vapor density   8.8 (vs air)
vapor pressure   1 mmHg ( 119 °C)
assay   ≥98.5% (GC)
autoignition temp.   455 °F
shelf life   (limited shelf life, expiry date on the label)
bp   317 °C(lit.)
mp   26-29 °C(lit.)
  28-30 °C
density   0.777 g/mL at 25 °C(lit.)

Price and Availability

Customers Also Viewed

Eicosane

analytical standard

Hexadecane

analytical standard

Octadecane

purum, ≥97.0% (GC)

Octadecane

99%

Octadecane

analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?