Synonym: (R)-(−)-3,7-Dimethyl-1,6-octadien-3-ol
CAS Number 126-91-0
Linear Formula (CH3)2C=CHCH2CH2C(CH3)(OH)CH=CH2
Molecular Weight 154.25
Beilstein Registry Number 1721487
EC Number 204-811-2
MDL number MFCD00135469
PubChem Substance ID 24886737
| Related Categories | Alcohols, Aliphatic, All Alphabetically Sorted, Allergenic Compounds, Alphabetical Index of Analytical Standards, |
| grade | analytical standard, for terpene analysis |
| assay | ≥98.0% (sum of enantiomers, GC) |
| optical activity | [α]20/D −20±2°, neat |
| shelf life | (limited shelf life, expiry date on the label) |
| refractive index |
n |
|
n | |
| bp | 198 °C(lit.) |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?