Synonym: 1-Octanesulfonic acid sodium salt monohydrate
CAS Number 207596-29-0
Linear Formula CH3(CH2)6CH2SO3Na · H2O
Molecular Weight 234.29
Beilstein Registry Number 3574241
EC Number 226-195-4
MDL number MFCD00149551
PubChem Substance ID 24886745
| Related Categories | Alkyl Sulfonates, Analytical Reagents, Analytical/Chromatography, Anionic Detergents, Basic Ion Pair Reagents (Cationic), |
| grade | for ion pair chromatography |
| description | anionic |
| assay | ≥99.0% (T) |
| mp | ≥300 °C(lit.) |
| λ | 10 % in H2O |
| UV absorption | λ: 210 nm Amax: 0.1 |
| λ: 220 nm Amax: 0.06 |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?