75075 Fluka

Sodium octyl sulfate

≥95.0% (S)

DOWNLOAD MSDS (PDF)

Synonym: Octyl sulfate sodium salt

  • CAS Number 142-31-4

  • Linear Formula CH3(CH2)7OSO3Na

  • Molecular Weight 232.27

  •  Beilstein Registry Number 3658635

  •  EC Number 205-535-5

  •  MDL number MFCD00007470

  • Popular Documents:  FTNMR (PDF)  

Properties

description   anionic
assay   ≥95.0% (S)
mp   195 °C (dec.)(lit.)

Price and Availability

Customers Also Viewed

Sodium octyl sulfate

for ion pair chromatography, ≥99.0% (T)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?