Synonym: Olivae oleum virginale
CAS Number 8001-25-0
MDL number MFCD00131764
Popular Documents: FTIR (PDF)
| Related Categories | Biochemicals and Reagents, Core Bioreagents, Lipids, Lipids and Related Products, Oils, |
| grade | Ph Eur |
| pharmacopeia | testing & handling conforms to Pharmacopeia |
The health benefits of olive oil result from its higher proportion of monounsaturated fats, as well as its antioxidant constituents: hydroxytyrosol, oleuropein and oleocanthal.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?