Synonym:
α-Naphthol orange,
4-
CAS Number 523-44-4
Empirical Formula (Hill Notation) C16H11N2NaO4S
Molecular Weight 350.32
Beilstein Registry Number 3826844
Colour Index Number 14600
EC Number 208-346-6
MDL number MFCD00021510
PubChem Substance ID 24886821
| Related Categories | Analytical Reagents, Analytical/Chromatography, Indicators, Microscopy Reagents, Stains and Dyes, |
| grade | for microscopy (Hist.) |
| indicator (pH 7.6-8.9) |
Staining of proteins1
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?