Synonym:
4-
CAS Number 633-96-5
Empirical Formula (Hill Notation) C16H11N2NaO4S
Molecular Weight 350.32
Beilstein Registry Number 3898201
Colour Index Number 15510
EC Number 211-199-0
MDL number MFCD00011657
PubChem Substance ID 24886822
| Related Categories | Analytical Reagents, Analytical/Chromatography, Microscopy Reagents, Stains and Dyes |
| grade | for microscopy (Hist.) |
| indicator (pH 11.0-13.0) |
Reagent for the extraction and spectrophotomeric determination of cationic surfactants1
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?