75563 Fluka

Ethyl butyrate

analytical standard

DOWNLOAD MSDS (PDF)

Synonym: Butyric acid ethyl ester

  • CAS Number 105-54-4

  • Linear Formula CH3CH2CH2C(O)OC2H5

  • Molecular Weight 116.16

  •  Beilstein Registry Number 506331

  •  EC Number 203-306-4

  •  MDL number MFCD00009394

  •  PubChem Substance ID 24886845

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories A-F, Alcoholic & Nonalcoholic Beverages, All Alphabetically Sorted, Alpha Sort, Alphabetical Index of Analytical Standards,
grade   analytical standard
vapor density   4 (vs air)
vapor pressure   15.5 mmHg ( 25 °C)
assay   ≥99.5% (GC)
autoignition temp.   865 °F
shelf life   (limited shelf life, expiry date on the label)
refractive index   n20/D 1.392(lit.)
  n20/D 1.393
bp   120 °C(lit.)
mp   −93 °C(lit.)
density   0.875 g/mL at 25 °C(lit.)

Price and Availability

Proficiency Testing Samples from RTC


PT Samples from RTCRTC has been producing environmental LPTP for more than 20 years and send out over 20,000 PT samples a year, throughout
the world.





Customers Also Viewed

Ethyl butyrate

≥98%, FCC

Ethyl butyrate

natural, ≥98%, FCC, FG

Ethyl hexanoate

≥99%

Ethyl propionate

analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?