EMAIL THIS PAGE TO A FRIEND

75805 Fluka

Oxford Agar

for microbiology

Synonym: Listeria Selective Agar, Oxford, Listeria Oxford Medium

Purchase

Properties

Related Categories All Dairy Media (alphabetical), All Food Control Media (alphabetical), All Solid Media/Agars, Listeria, Media,
grade   for microbiology
shelf life   limited shelf life, expiry date on the label
composition   agar, 10 g/L
  corn starch, 1 g/L
  esculin, 1 g/L
  ferric ammonium citrate, 0.5 g/L
  lithium chloride, 15 g/L
  sodium chloride, 5 g/L
  special peptone, 23 g/L
final pH   7.0±0.2 (25 °C)
suitability   selective and differential for Listeria

Description

Application

For the isolation and cultivation of Listeria monocytogenes from specimens containing a mixed bacterial flora.

Other Notes

Selective differential medium for the isolation of L. monocytogenes1

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Hazard Codes (Europe) 
Xn
Risk Statements (Europe) 
Safety Statements (Europe) 
26
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Listeria a Survivor

Listeriosis is a serious infection caused by Listeria monocytogenes. In recent years it has been recognized that Listeria is an important public health problem. The disease affects primarily people o...
Jvo Siegrist
Microbiology Focus Edition 2.2
Keywords: AGE, Antibiotics, Diseases, Fermentation, Food Safety, Gastrointestinal, Indicators, Methods, Microbiology, Nutrition, Purification

Peer-Reviewed Papers

References

Set your institution to view full text papers.

1. Curtis, G.D.W., et al. Lett. Appl. Microbiol. 8, 95, (1989)

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?