Synonym: 2-Oxoglutaric acid, 2-Oxopentanedioic acid, alpha-Ketoglutaric acid
CAS Number 328-50-7
Linear Formula HOOCCH2CH2COCOOH
Molecular Weight 146.10
Beilstein Registry Number 1705689
EC Number 206-330-3
MDL number MFCD00004165
Popular Documents: FTNMR (PDF)
| Related Categories | Building Blocks, C1 to C5, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis, |
| Quality Level | PREMIUM |
| assay | ≥99.0% (T) |
| ign. residue | ≤0.05% |
| mp | 114-117 °C |
| solubility | H2O: soluble0.1 g/mL, clear, colorless |
| storage temp. | 2-8°C |
Reagent for the analysis of tryptophan1; Substrate for glutamate dehydrogenase
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?