Synonym:
1-
CAS Number 57-11-4
Linear Formula CH3(CH2)16COOH
Molecular Weight 284.48
Beilstein Registry Number 608585
E Number E570
EC Number 200-313-4
MDL number MFCD00002752
| Related Categories | All Alphabetically Sorted, Analytical Standards, Analytical/Chromatography, CRM for chromatography, Certified Reference Materials (CRMs), |
| grade | certified reference material |
| vapor pressure | 1 mmHg ( 173.7 °C) |
| grade | TraceCERT® |
| shelf life | (limited shelf life, expiry date on the label) |
| bp | 361 °C(lit.) |
| mp | 67-72 °C(lit.) |
Certified content by quantitative NMR incl. uncertainty and expiry are given in the certificate.
Download your certificate at: http://www.sigma-aldrich.
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?