Synonym:
4-Amino-N-
CAS Number 723-46-6
Empirical Formula (Hill Notation) C10H11N3O3S
Molecular Weight 253.28
Beilstein Registry Number 6732984
EC Number 211-963-3
MDL number MFCD00010546
| Related Categories | APIs (Active Pharmaceutical Ingredients) & Metabolites, All alphabetically sorted, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, |
| grade | certified reference material |
| TraceCERT® | |
| shelf life | (limited shelf life, expiry date on the label) |
Certified content by quantitative NMR incl. uncertainty and expiry are given in the certificate.
Download your certificate at: http://www.sigma-aldrich.
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC
Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?