Synonym: Paraffinum solidum
CAS Number 8002-74-2
EC Number 232-315-6
MDL number MFCD00132833
Popular Documents: FTIR (PDF)
| Related Categories | Core Bioreagents, Lipids and Related Products, Pharmacopoeia, Pharmacopoeia A-Z, Research Essentials More... |
| grade | Ph Eur |
| form | pellets |
| pharmacopeia | testing & handling conforms to Pharmacopeia |
| color | white |
Customers Also Viewed
mp 53-57 °C (ASTM D 87)
mp 58-62 °C (ASTM D 87)
meets analytical specification of Ph. Eur., white, pastilles
pellets, white
pellets, white
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?