Synonym: Methyl pelargonate, Nonanoic acid methyl ester
CAS Number 1731-84-6
Linear Formula CH3(CH2)7COOCH3
Molecular Weight 172.26
Beilstein Registry Number 1754521
EC Number 217-052-7
MDL number MFCD00009569
PubChem Substance ID 24886979
Popular Documents: FTNMR (PDF)
| Related Categories | Alcoholic & Nonalcoholic Beverages, All Alphabetically Sorted, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, |
| grade | analytical standard |
| assay | ≥99.8% (GC) |
| shelf life | (limited shelf life, expiry date on the label) |
| refractive index |
n |
|
n | |
| bp | 213-214 °C(lit.) |
| density | 0.875 g/mL at 25 °C(lit.) |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?