• USA Home
  • 76828 - N,N,N′,N′′,N′′-Pentamethyldiethylenetriamine

EMAIL THIS PAGE TO A FRIEND

76828 Fluka

N,N,N′,N′′,N′′-Pentamethyldiethylenetriamine

purum, ≥98.0% (GC)

Synonym: PMDETA, PMDTA

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Nitrogen Compounds, Organic Building Blocks, Polyamines More...
vapor pressure   0.23 mmHg ( 20 °C)
grade   purum
assay   ≥98.0% (GC)
autoignition temp.   311 °F
expl. lim.   5.6 %
impurities   ≤0.5% water
refractive index   n20/D 1.442(lit.)
  n20/D 1.443
bp   198 °C(lit.)
mp   −20 °C(lit.)
density   0.83 g/mL at 25 °C(lit.)

Description

Other Notes

Catalyst component for Ni(II)-mediated alkyne carboxylation1

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Hazard Codes (Europe) 
T
Risk Statements (Europe) 
Safety Statements (Europe) 
26-36/37/39-45
RIDADR 
UN 2734PSN2 3(8) / PGII
WGK Germany 
2
Flash Point(F) 
127.4 °F
Flash Point(C) 
53 °C

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

1. E. Dunach, J. Périchon Synlett, 143, (1990)

Beil. 4,IV,1240

FT-NMR 1 (1), 499:C / RegBook 1 (1), 331:B / Structure Index 1, 45:B:8

Related Products

replaced by

Product #

Description

Add to Cart

369497 N,N,N′,N′′,N′′-Pentamethyldiethylenetriamine, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?