77239 Fluka

Sulfuric acid

≥95%, TraceSELECT® Ultra, for ultratrace analysis

DOWNLOAD MSDS (PDF)
  • CAS Number 7664-93-9

  • Linear Formula H2SO4

  • Molecular Weight 98.08

  •  Beilstein Registry Number 2037554

  •  EC Number 231-639-5

  •  MDL number MFCD00064589

  •  PubChem Substance ID 24886817

Properties

Related Categories Acids & Bases, Analytical Reagents, Analytical/Chromatography, Atomic Absorption Spectroscopy (AAS), Digestion Reagents,
grade   for ultratrace analysis
vapor density   <0.3 (25 °C, vs air)
vapor pressure   1 mmHg ( 146 °C)
description   Nominally 95-98% H2SO4
grade   TraceSELECT® Ultra
assay   ≥95% (T)
  ≥95%
bp   ~290 °C(lit.)
density   1.83 g/mL at 20 °C
  1.840 g/mL at 25 °C(lit.)
anion traces   chloride (Cl-): ≤0.5 mg/kg
  nitrate (NO3-): ≤0.1 mg/kg
  phosphate (PO43-): ≤0.5 mg/kg
cation traces   Ag: ≤0.01 μg/kg
  Al: ≤0.5 μg/kg
  As: ≤1 μg/kg
  Au: ≤0.02 μg/kg
  Ba: ≤0.02 μg/kg
  Be: ≤0.01 μg/kg
  Bi: ≤0.01 μg/kg
  Ca: ≤2 μg/kg
  Cd: ≤0.01 μg/kg
  Ce: ≤0.01 μg/kg
  Co: ≤0.01 μg/kg
  Cr: ≤0.05 μg/kg
  Cs: ≤0.01 μg/kg
  Cu: ≤0.5 μg/kg
  Dy: ≤0.01 μg/kg
  Er: ≤0.01 μg/kg
  Eu: ≤0.01 μg/kg
  Fe: ≤0.5 μg/kg
  Ga: ≤0.01 μg/kg
  Gd: ≤0.01 μg/kg
  Ge: ≤0.01 μg/kg
  Hf: ≤0.01 μg/kg
  Hg: ≤0.5 μg/kg
  Ho: ≤0.01 μg/kg
  In: ≤0.01 μg/kg
  Ir: ≤0.01 μg/kg
  K: ≤0.5 μg/kg
  La: ≤0.01 μg/kg
  Li: ≤0.02 μg/kg
  Lu: ≤0.01 μg/kg
  Mg: ≤0.2 μg/kg
  Mn: ≤0.02 μg/kg
  Mo: ≤0.05 μg/kg
  Na: ≤1 μg/kg
  Nb: ≤0.01 μg/kg
  Nd: ≤0.01 μg/kg
  Ni: ≤0.1 μg/kg
  Pb: ≤0.05 μg/kg
  Pd: ≤0.01 μg/kg
  Pr: ≤0.01 μg/kg
  Pt: ≤0.01 mg/kg
  Rb: ≤0.01 μg/kg
  Rh: ≤0.01 μg/kg
  Sb: ≤0.05 μg/kg
  Sc: ≤0.02 μg/kg
  Se: ≤1 μg/kg
  Sm: ≤0.01 μg/kg
  Sn: ≤0.5 μg/kg
  Sr: ≤0.01 μg/kg
  Tb: ≤0.01 μg/kg
  Te: ≤0.01 μg/kg
  Th: ≤0.01 μg/kg
  Ti: ≤0.1 μg/kg
  Tl: ≤0.01 μg/kg
  Tm: ≤0.01 μg/kg
  U: ≤0.01 μg/kg
  V: ≤0.01 μg/kg
  W: ≤0.01 μg/kg
  Y: ≤0.01 μg/kg
  Yb: ≤0.01 μg/kg
  Zn: ≤0.5 μg/kg
  Zr: ≤0.02 μg/kg

Description

Legal Information

TraceSELECT is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Customers Also Viewed

Acetic acid

ACS reagent, ≥99.7%

Sulfuric acid

99.999%

Sulfuric acid

≥95%, TraceSELECT®, for trace analysis

Sulfuric acid

ACS reagent, 95.0-98.0%

Sulfuric acid

ACS reagent, 95.0-98.0%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?