• USA Home
  • 77247 - Heptafluorobutyric acid

EMAIL THIS PAGE TO A FRIEND

77247 Fluka

Heptafluorobutyric acid

≥99.0% (GC)

Synonym: Edman Reagent No. 3, HFBA, Perfluorobutyric acid

Purchase

Properties

Related Categories Molecular Biology, Protein Sequencing, Protein Structural Analysis, Proteomics, Reagents for Protein Sequencing More...
vapor density   7 (vs air)
vapor pressure   ~10 mmHg ( 25 °C)
assay   ≥99.0% (GC)
refractive index   n20/D 1.3(lit.)
bp   120 °C/755 mmHg(lit.)
density   1.645 g/mL at 25 °C(lit.)

Description

Other Notes

This product has been replaced by 77249-SIGMA | Heptafluorobutyric acid ≥99.0% (GC)

Price and Availability

TruPAGE™ Pre-cast Polyacrylamide Gels


TruPAGE™ Pre-cast Polyacrylamide GelsTruPAGE Gels run true every time!

  • . 2-Year Shelf Life
  • . Superior Band Resolution
  • . Strong & Tear Resistant
  • . Higher Capacity Wells



Duolink Webinar

Customers Also Viewed

Heptafluorobutyric acid

≥99.0% (GC)

Heptafluorobutyric acid

for ion chromatography, ≥99.5% (GC)

Heptafluorobutyric acid solution

0.5 M in H2O, for ion pair chromatography

Heptafluorobutyric anhydride

for GC derivatization, ≥99.0%

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
Hazard Codes (Europe) 
C
Risk Statements (Europe) 
35
Safety Statements (Europe) 
26-36/37/39-45
RIDADR 
UN 3265 8 / PGI
WGK Germany 
3
RTECS 
ET4025000

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Distribution and sources of polyfluoroalkyl substances (PFAS) in the River Rhine watershed. Möller A, Ahrens L, Surm R, et al. Environ. Pollut. 158(10), 3243-50, (2010)

Loading...


Toxicological evaluation of ammonium perfluorobutyrate in rats: twenty-eight-day and ninety-day oral gavage studies. Butenhoff JL, Bjork JA, Chang SC, et al. Reprod. Toxicol. 33(4), 513-30, (2012)

Loading...


Differential hepatic effects of perfluorobutyrate mediated by mouse and human PPAR-alpha. Foreman JE, Chang SC, Ehresman DJ, et al. Toxicol. Sci. 110(1), 204-11, (2009)

Loading...


Structure-activity relationship assessment of four perfluorinated chemicals using a prolonged zebrafish early life stage test. Hagenaars A, Vergauwen L, De Coen W, et al. Chemosphere 82(5), 764-72, (2011)

Loading...


Cooperative effect in ion pairing of oligolysine with heptafluorobutyric acid in reversed-phase chromatography. Xie W, Qin X, Teraoka I, et al. J. Chromatogr. A 1218(43), 7765-70, (2011)

Loading...


Distribution and primary source analysis of per- and poly-fluoroalkyl substances with different chain lengths in surface and groundwater in two cities, North China. Yao Y, Zhu H, Li B, et al. Ecotoxicol. Environ. Saf. 108, 318-28, (2014)

Loading...


Analysis of perfluoroalkyl substances in waters from Germany and Spain. Llorca M, Farré M, Picó Y, et al. Sci. Total Environ. 431, 139-50, (2012)

Loading...


Structural analysis of naturally occurring sialic acids. Kamerling JP and Gerwig GJ Methods Mol. Biol. 347, 69-91, (2006)

Loading...


Polyfluoroalkyl compounds in landfill leachates. Busch J, Ahrens L, Sturm R, et al. Environ. Pollut. 158(5), 1467-71, (2010)

Loading...


Comparative pharmacokinetics of perfluorobutyrate in rats, mice, monkeys, and humans and relevance to human exposure via drinking water. Chang SC, Das K, Ehresman DJ, et al. Toxicol. Sci. 104(1), 40-53, (2008)

Loading...


Light-induced degradation of perfluorocarboxylic acids in the presence of titanium dioxide. Dillert R, Bahnemann D, and Hidaka H Chemosphere 67(4), 785-92, (2007)

Loading...


Effects of perfluorobutyrate exposure during pregnancy in the mouse. Das KP, Grey BE, Zehr RD, et al. Toxicol. Sci. 105(1), 173-81, (2008)

Loading...


Behavior of perfluorinated compounds in soils during leaching experiments. Gellrich V, Stahl T, and Knepper TP Chemosphere 87(9), 1052-6, (2012)

Loading...


Application of perfluorinated acids as ion-pairing reagents for reversed-phase chromatography and retention-hydrophobicity relationships studies of selected beta-blockers. Flieger J J. Chromatogr. A 1217(4), 540-9, (2010)

Loading...


Perfluoroalkyl substances (PFASs) in food and water from Faroe Islands. Eriksson U, Kärrman A, Rotander A, et al. Environ. Sci. Pollut. Res. Int. 20(11), 7940-8, (2013)

Loading...


Simultaneous detection and quantitation of highly water-soluble herbicides in serum using ion-pair liquid chromatography-tandem mass spectrometry. Wang KC, Chen SM, Hsu JF, et al. J. Chromatogr. B. Analyt. Technol. Biomed. Life Sci. 876(2), 211-8, (2008)

Loading...


Analysis of underivatized polyamines by reversed phase liquid chromatography with electrospray tandem mass spectrometry. Häkkinen MR, Keinänen TA, Vepsäläinen J, et al. J. Pharm. Biomed. Anal. 45(4), 625-34, (2007)

Loading...


Solvolytic reactivity of heptafluorobutyrates and trifluoroacetates. Denegri B and Kronja O J. Org. Chem. 74(16), 5927-33, (2009)

Loading...


Separation of polar betalain pigments from cacti fruits of Hylocereus polyrhizus by ion-pair high-speed countercurrent chromatography. Wybraniec S, Stalica P, Jerz G, et al. J. Chromatogr. A 1216(41), 6890-9, (2009)

Loading...


Simultaneous determination of flavonoid and alkaloid compounds in Citrus herbs by high-performance liquid chromatography-photodiode array detection-electrospray mass spectrometry. Ding L, Luo X, Tang F, et al. J. Chromatogr. B. Analyt. Technol. Biomed. Life Sci. 857(2), 202-9, (2007)

Loading...


Adsorption of four perfluorinated acids on non ion exchange polymer sorbents. Senevirathna ST, Tanaka S, Fujii S, et al. Water Sci. Technol. 63(10), 2106-13, (2011)

Loading...


Separation of Amadori peptides from their unmodified analogs by ion-pairing RP-HPLC with heptafluorobutyric acid as ion-pair reagent. Frolov A and Hoffmann R Anal. Bioanal. Chem 392(6), 1209-14, (2008)

Loading...


GC separation of amino acid enantiomers via derivatization with heptafluorobutyl chloroformate and Chirasil-L-Val column. Zahradnícková H, Husek P, and Simek P J. Sep. Sci. 32(22), 3919-24, (2009)

Loading...


Underivatized polyamine analysis in plant samples by ion pair LC coupled with electrospray tandem mass spectrometry. Sánchez-López J, Camañes G, Flors V, et al. Plant Physiol. Biochem. 47(7), 592-8, (2009)

Loading...


Quantitation of quinapril in human plasma by matrix-assisted laser desorption ionization time-of-flight mass spectrometry with quinolone matrix additives. Lu CY, Liu FT, and Feng CH J. Chromatogr. B. Analyt. Technol. Biomed. Life Sci. 879(26), 2688-94, (2011)

Loading...


Accumulation of perfluoroalkyl substances in human tissues. Pérez F, Nadal M, Navarro-Ortega A, et al. Environ. Int. 59, 354-62, (2013)

Loading...


Preconcentration and decomposition of perfluorinated carboxylic acids on an activated charcoal cartridge with sodium biphenyl reagent and its determination at microg L(-1) level on the basis of flow injection-fluorimetric detection of fluoride ion. Takayanagi T, Yamashita H, Motomizu S, et al. Talanta 74(5), 1224-30, (2008)

Loading...


Separation of amaranthine-type betacyanins by ion-pair high-speed countercurrent chromatography. Jerz G, Gebers N, Szot D, et al. J. Chromatogr. A 1344, 42-50, (2014)

Loading...


Interconversion of ephedrine and pseudoephedrine during chemical derivatization. Wong CH, Ho EN, Kwok WH, et al. Drug Test. Anal. 4(12), 1028-33, (2012)

Loading...


Beil. 2,IV,810

Aldrich MSDS 1, 1005:A / Corp MSDS 1 (1), 1819:A / FT-IR 2 (1), 796:D / FT-IR 1 (1), 507:A / FT-NMR 1 (1), 800:B / IR-Spectra (3), 297:E / IR-Spectra (2), 268:B / RegBook 1 (1), 559:J / Sax 6, 1498 / Sigma FT-IR 1 (2), 602:A / Structure Index 1, 79:D:7 / Vapor Phase 3, 590:A

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?