77624 Fluka

Dexpanthenol

Ph Eur

DOWNLOAD MSDS (PDF)

Synonym: (R)-(+)-2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutyramide, (R)-2,4-Dihydroxy-3,3-dimethylbutyric 3-hydroxypropylamide, D-Panthenol, D-Pantothenyl alcohol, Dexpanthenol, Dexpanthenolum, Provitamin B

  • CAS Number 81-13-0

  • Empirical Formula (Hill Notation) C9H19NO4

  • Molecular Weight 205.25

  •  Beilstein Registry Number 1724947

  •  EC Number 201-327-3

  •  MDL number MFCD00065006

  •  PubChem Substance ID 24887211

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Core Bioreagents, Organic Building Blocks,
grade   Ph Eur
pharmacopeia   testing & handling conforms to Pharmacopeia
density   1.20 g/mL at 20 °C(lit.)

Price and Availability

Customers Also Viewed

(±)-1,3-Butanediol

puriss., ≥99.0% (GC)

Calcium pantothenate

meets USP testing specifications, monograph mol wt. 476.53 (C18H32CaN2O10)

<SC>D</SC>-Panthenol

≥98% (perchloric acid titration), ≥98% (TLC)

<SC>D</SC>-Panthenol

≥98.0% (NT)

<SC>DL</SC>-Panthenol

99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?