• USA Home
  • 77889 - 3-[3-Hydroxy-4-(5-nitro-2-pyridylazo)-N-propylanilino]propanesulfonic acid disodium salt dihydrate

EMAIL THIS PAGE TO A FRIEND

77889 Fluka

3-[3-Hydroxy-4-(5-nitro-2-pyridylazo)-N-propylanilino]propanesulfonic acid disodium salt dihydrate

purum, ≥97.0% (HPLC)

Synonym: 2-(5-Nitro-2-pyridylazo)-5-(N-propyl-3-sulfopropylamino)phenol disodium salt, Nitro-PAPS

  • CAS Number 143205-66-7

  • Empirical Formula (Hill Notation) C17H19N5Na2O6S · 2H2O

  • Molecular Weight 503.44

  •  MDL number MFCD00037623

  •  PubChem Substance ID 24887142

Purchase

Properties

grade   purum
assay   ≥97.0% (HPLC)
impurities   ~10% water

Description

Other Notes

nitro-PAPS is a chromogenic reagent for the colorimetric assay of serum iron and zinc1,2; assay of micro amounts of metals following ion-pair extn. with nitro-PAPS3,4

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Price and Availability

Customers Also Viewed

2-<WBR>(5-<WBR>Bromo-<WBR>2-<WBR>pyridylazo)<WBR>-<WBR>5-<WBR>[<I>N</I>-propyl-<I>N</I>-<WBR>(3-<WBR>sulfopropyl)<WBR>amino]<WBR>phenol disodium salt dihydrate

for spectrophotometric det. of Zn(II), Cu(II), Fe(II), Co(II), H2O2, ≥95.0%

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?