77981 Fluka

CT Supplement

for microbiology

DOWNLOAD MSDS (PDF)

Synonym: Cefixim Tellurite Supplement, Tellurite Cefixim Supplement

Properties

Related Categories Media, Microbiology, Supplements More...
grade   for microbiology
shelf life   (limited shelf life, expiry date on the label)

Description

Components

(one vial sufficient for 500 mL medium) Potassium tellurite 1.25 mg, Cefixime 0.025 mg

Application

A selective supplement recommended by ISO Committee for the isolation of Escherichia coli 0157.

Price and Availability

Customers Also Viewed

Cefixime trihydrate

≥98.0% (HPLC)

Potassium tellurite hydrate

≥95.0% (calc. on dry substance, RT)

Potassium tellurite solution

1% in H2O, for microbiology

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?