Synonym:
Quercetin-
CAS Number 250249-75-3
Empirical Formula (Hill Notation) C27H30O16 · 3H2O
Molecular Weight 664.56
Beilstein Registry Number 75455
EC Number 205-814-1
MDL number MFCD00149490
PubChem Substance ID 24887256
| Related Categories | Acacia, Alcohols, Allium cepa (Onion), Allium sativum (Garlic), Alphabetical Index of Analytical Standards, |
| grade | analytical standard |
| assay | ≥95.0% (HPLC) |
| optical activity | [α]20/D +9±2°, c = 0.5% in absolute ethanol |
Customers Also Viewed
≥95% (HPLC), solid
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?