Synonym:
2-
CAS Number 156-06-9
Linear Formula C6H5CH2COCOOH
Molecular Weight 164.16
Beilstein Registry Number 2207312
EC Number 205-847-1
MDL number MFCD00002589
Popular Documents: FTNMR (PDF)
| Related Categories | Building Blocks, C9, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis, |
| grade | purum |
| assay | ≥98.0% (T) |
| mp | 150-154 °C(lit.) |
| 150-154 °C | |
| storage temp. | −20°C |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?