78195 Fluka

Calcium phenylpyruvate

purum, ≥98.0% (dry substance, NT)

DOWNLOAD MSDS (PDF)

Synonym: Phenylpyruvic acid calcium salt

  • CAS Number 51828-93-4

  • Linear Formula (C6H5CH2COCOO)2Ca

  • Molecular Weight 366.38

  •  EC Number 257-455-5

  •  MDL number MFCD00037200

  •  PubChem Substance ID 24887273

Properties

Related Categories Building Blocks, Carbonyl Compounds, Carboxylic Acid Salts, Chemical Synthesis, Organic Building Blocks More...
grade   purum
assay   ≥98.0% (dry substance, NT)
impurities   ≤6% water

Price and Availability

Customers Also Viewed

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?