• USA Home
  • 79311 - Malvidin-3-galactoside chloride

EMAIL THIS PAGE TO A FRIEND

79311 Fluka

Malvidin-3-galactoside chloride

analytical standard

Synonym: Primulin

Purchase

Price and Availability

Customers Also Viewed

Malvidin chloride

analytical standard

Malvidin chloride

≥95.0% (HPLC)

Malvin(chloride)

≥90% (HPLC)

Oenin chloride

≥90% (HPLC)

Peonidin 3-<I>O</I>-glucoside chloride

analytical standard

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Isolation of two developmentally regulated genes involved in spore wall maturation in Saccharomyces cerevisiae. Briza P, Breitenbach M, Ellinger A, et al. Genes Dev. 4(10), 1775-89, (1990)

Loading...


Absorption of anthocyanins from blueberries and serum antioxidant status in human subjects. Mazza G, Kay CD, Cottrell T, et al. J. Agric. Food Chem. 50(26), 7731-7, (2002)

Loading...


Bog bilberry (Vaccinium uliginosum L.) extract reduces cultured Hep-G2, Caco-2, and 3T3-L1 cell viability, affects cell cycle progression, and has variable effects on membrane permeability. Liu J, Zhang W, Jing H, et al. J. Food Sci. 75(3), H103-7, (2010)

Loading...


[Study of interneuronal connections by the technic of luminescent demonstration of retrograde transport of primulin]. Otellin VA, Meshkenaĭte VI, and Rybakov VL Arkh. Anat. Gistol. Embriol. 80(1), 26-9, (1981)

Loading...


Initial neurons of associative and callosal pathways of lateral suprasylvian area of the cat brain. Otellin VA and Meshkenaite VI Neurosci. Behav. Physiol. 16(5), 441-7, (1986)

Loading...


[Role of membrane damage in the inactivation of yeast cells by visible light]. Pospelov ME, Turovetskiĭ VB, and Fraĭkin GIa Mikrobiologiia 56(5), 882-5, (1987)

Loading...


[Connections between neurons of sympathetic ganglia and the myenteric nerve plexus of the mammalian colon]. Riakhovskaia LV and Adamatskiĭ AI Arkh. Anat. Gistol. Embriol. 91(10), 15-21, (1986)

Loading...


[The sources of the innervation of the sphincter of the esophagogastric junction in rats]. Kolesnikov LL Arkh. Anat. Gistol. Embriol. 100(5), 28-37, (1991)

Loading...


[The structure of the initial inputs into the metasympathetic nervous system of the rat uterus]. Kucheriavykh LE, Skopichev VG, and Nozdrachev AD Morfologiia. 116(6), 22-5, (1999)

Loading...


Phenolic compounds and bioactivities of pigmented rice. Deng GF, Xu XR, Zhang Y, et al. Crit. Rev. Food Sci. Nutr 53(3), 296-306, (2013)

Loading...


High-resolution separation and quantification of neutral lipid and phospholipid species in mammalian cells and sera by multi-one-dimensional thin-layer chromatography. White T, Bursten S, Federighi D, et al. Anal. Biochem. 258(1), 109-17, (1998)

Loading...


Divergent projections of catecholamine neurons of the locus coeruleus as revealed by fluorescent retrograde double labeling technique. Nagai T, Satoh K, Imamoto K, et al. Neurosci. Lett. 23(2), 117-23, (1981)

Loading...


[Intravital study of the hydrophobic interactions in protein fibrils by a polarization-fluorescence method. I. Collagen fibrils]. Gamaleĭ IA and Kaulin AB Tsitologiia 29(12), 1365-71, (1987)

Loading...


[Identification of catecholamines in the rat brain and mollusk tissues using semithin slices and retrograde staining]. Prudnikov IM, Maĭskiĭ VA, and Chepera EN Fiziol. Zh. 35(6), 98-100, (1989)

Loading...


Characterisation of minor tetra- to hepta-saccharides O-linked to human meconium glycoproteins by t.l.c.-m.s. microsequencing of neoglycolipid derivatives in conjunction with conventional m.s. and 1H-n.m.r. spectroscopy. Lawson AM, Hounsell EF, Stoll MS, et al. Carbohydr. Res. 221, 191-208, (1991)

Loading...


Direct effects of Vaccinium myrtillus L. fruit extracts on rat heart mitochondrial functions. Trumbeckaitė S, Burdulis D, Raudonė L, et al. Phytother Res. 27(4), 499-506, (2013)

Loading...


Retrograde labeling and dissociation of mouse retinal ganglion cells. Sarthy PV and Balkema GW Neurosci. Lett. 25(3), 205-8, (1981)

Loading...


Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?