• USA Home
  • 79560 - Phosphomolybdic acid hydrate

EMAIL THIS PAGE TO A FRIEND

79560 Fluka

Phosphomolybdic acid hydrate

for microscopy

Synonym: Molybdophosphoric acid

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Chemical Synthesis, Microscopy Reagents, Others,
grade   for microscopy
loss   21-23% loss on drying, 110 °C
density   1.62 g/mL at 25 °C(lit.)
anion traces   chloride (Cl-): ≤50 mg/kg
  sulfate (SO42-): ≤100 mg/kg
cation traces   Ca: ≤50 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤20 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤50 mg/kg
  K: ≤50 mg/kg
  Mg: ≤20 mg/kg
  Mn: ≤100 mg/kg
  NH4+: ≤30 mg/kg
  Na: ≤50 mg/kg
  Ni: ≤10 mg/kg
  Pb: ≤20 mg/kg
  Zn: ≤50 mg/kg

Description

Application

A precursor to hybrid organic-inorganic based molecular batteries with applications as cation-insertion electrodes.1

Other Notes

Review: of the folin phenol method for the quantitative determination of proteins2

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Customers Also Viewed

Phosphomolybdic acid hydrate

≥99.99% trace metals basis

Phosphomolybdic acid hydrate

moist crystals

Phosphomolybdic acid solution

ready-to-use spray and plunge reagent, for chromatography

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Hazard Codes (Europe) 
O,C
Risk Statements (Europe) 
Safety Statements (Europe) 
17-26-36/37/39-45
RIDADR 
UN 3084 5.1(8) / PGII
WGK Germany 
3

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

1. Lira-Cantú, M.; Gómez-Romero, P. Chem. Mater. 10, 698, (1988)

2. Review of the Folin phenol protein quantitation method of Lowry, Rosebrough, Farr and Randall. G.L. Peterson Anal. Biochem. 100, 201, (1979)

Loading...


Ultra-deep oxidative desulfurization of diesel fuel by the Mo/Al 2 O 3-H 2 O 2 system: The effect of system parameters on catalytic activity. Garcia-Gutierrez, Jose Luis, et al. Appl. Catal. A Gen. 334, 366-373, (2008)

Chemical News A new precipitant for Potash, (1868), 183

Corp MSDS 1 (2), 2820:B / RegBook 1 (3), 3255:B

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?