79690 Fluka

Phosphotungstic acid hydrate

for microscopy

DOWNLOAD MSDS (PDF)

Synonym: Tungstophosphoric acid

  • CAS Number 12501-23-4

  • Linear Formula H3[P(W3O10)4] · xH2O

  • Molecular Weight 2880.05 (anhydrous basis)

  •  MDL number MFCD00149915

  •  PubChem Substance ID 24887496

Properties

Related Categories Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, Derivatization Reagents, Derivatization Reagents TLC,
grade   for microscopy
impurities   ≤0.002% total nitrogen (N)
loss   ≤15% loss on drying
mp   ~95 °C(lit.)
anion traces   chloride (Cl-): ≤50 mg/kg
  sulfate (SO42-): ≤100 mg/kg
cation traces   Ca: ≤20 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤20 mg/kg
  K: ≤50 mg/kg
  Na: ≤50 mg/kg
  Pb: ≤20 mg/kg
  Zn: ≤5 mg/kg

Description

Other Notes

Review of the folin phenol method for the quantitative determination of proteins1

Price and Availability

Customers Also Viewed

Phosphomolybdic acid hydrate

for microscopy

Phosphotungstic acid hydrate

99.995% trace metals basis (Purity excludes up to 300 ppm Si)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?