80135 Fluka

Diisodecyl phthalate

Selectophore, ≥99.0%

DOWNLOAD MSDS (PDF)
  • CAS Number 26761-40-0

  • Empirical Formula (Hill Notation) C28H46O4

  • Molecular Weight 446.66

  •  Beilstein Registry Number 2171889

  •  EC Number 247-977-1

  •  MDL number MFCD00048389

  •  PubChem Substance ID 24887527

Properties

Related Categories Analytical/Chromatography, Ion Sensor Materials, Plasticizer for ISE More...
grade   Selectophore
assay   ≥99.0% (sum of isomers, GC)
  ≥99.0%
refractive index   n20/D 1.486
density   0.965 g/mL at 20 °C(lit.)

Description

Application

Plasticizer for PVC membrane electrodes

General description

Visit our Sensor Applications portal to learn more.

Legal Information

Selectophore is a trademark of Sigma-Aldrich GmbH

Price and Availability

Customers Also Viewed

Bis(2-ethylhexyl) phthalate

PESTANAL®, analytical standard

Di-n-octyl phthalate

analytical standard, for environmental analysis

Di-n-octyl phthalate

≥98.0% (GC)

Diisononyl phthalate

ester content ≥99 % (mixture of C9 isomers), technical grade

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?