80154 Fluka

Dipentyl phthalate

Selectophore, ≥99.0%

DOWNLOAD MSDS (PDF)
  • CAS Number 131-18-0

  • Empirical Formula (Hill Notation) C18H26O4

  • Molecular Weight 306.40

  •  Beilstein Registry Number 1987323

  •  EC Number 205-017-9

  •  MDL number MFCD00041934

  •  PubChem Substance ID 24887532

Properties

Related Categories Analytical/Chromatography, Ion Sensor Materials, Plasticizer for ISE More...
grade   Selectophore
assay   ≥99.0% (GC)
  ≥99.0%
refractive index   n20/D 1.490
density   1.025 g/mL at 20 °C(lit.)

Description

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Plasticizer for PVC membrane electrodes1,2

Legal Information

Selectophore is a trademark of Sigma-Aldrich GmbH

Price and Availability

Customers Also Viewed

Bis(2-ethylhexyl) phthalate

PESTANAL®, analytical standard

Bis(methylglycol) phthalate

analytical standard, for environmental analysis

Dioctyl phthalate

≥99.5%

Dipentyl phthalate

analytical standard

Dipentyl phthalate-3,4,5,6-d<SUB>4</SUB>

98 atom % D, 99% (CP)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?