80180 Fluka

Phytic acid solution

technical, ~40% in H2O

DOWNLOAD MSDS (PDF)

Synonym: myo-Inositol hexakis(dihydrogen phosphate)

  • CAS Number 83-86-3

  • Empirical Formula (Hill Notation) C6H18O24P6

  • Molecular Weight 660.04

  •  Beilstein Registry Number 2201952

  •  MDL number MFCD00082309

  •  PubChem Substance ID 24887537

Properties

Related Categories Carbohydrate Chemistry, Carbohydrate Derivatives, Chemical Biology, Chemical Synthesis
grade   technical
concentration   ~40% in H2O
refractive index   n20/D 1.39

Description

Application

Complexing agent for the removal of traces of heavy metal ions

Packaging

50, 250 mL in glass bottle

Other Notes

Tandem Mass Spectrometry data independently generated by Scripps Center for Metabolomics is available to view or download in PDF. 80180.pdf Tested metabolites are featured on Scripps Center for Metabolomics METLIN Metabolite Database. To learn more, visit sigma.com/metlin.

Price and Availability

Customers Also Viewed

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?