EMAIL THIS PAGE TO A FRIEND

80353 Fluka

TDA Reagent

for microbiology

Synonym: Ferric chloride solution, Iron(III) chloride solution, Tryptophan Deaminase Reagent

Purchase

Properties

Related Categories Biochemical Reagents for Identification, Identification Tests & Reagents, Microbiology More...
grade   for microbiology
shelf life   limited shelf life, expiry date on the label
composition   ferric chloride, 1 g/10 mL (in distilled water)

Description

Application

For identification of Proteus species by detection of tryptophan deaminase activity.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Customers Also Viewed

for microbiology

for microbiology

DMACA Reagent

for microbiology

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Hazard Codes (Europe) 
Xi
Risk Statements (Europe) 
41
Safety Statements (Europe) 
26-39
RIDADR 
UN 2582 8 / PGIII
WGK Germany 
1

Protocols & Articles

Articles

Identification of Microorganisms Based on Color

For microbiologists the most fundamental stain was developed in 1884 by the Danish bacteriologist Hans Christian Gram. The Gram Stain allows the determination of morphology, dividing bacteria into tw...
Jvo Siegrist
Microbiology Focus Edition 1.4, Product Manager Microbiology.... ivo.siegrist@sial.com
Keywords: Antibiotics, Enzyme activity, Fermentation, Gram stain, Help, Indicators, Reductions

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Oxidation inhibits iron-induced blood coagulation. Pretorius E, Bester J, Vermeulen N, et al. Curr. Drug Targets 14(1), 13-9, (2013)

Loading...


Tuning the reactivity of TEMPO by coordination to a Lewis acid: isolation and reactivity of MCl3(η1-TEMPO) (M = Fe, Al). Scepaniak JJ, Wright AM, Lewis RA, et al. J. Am. Chem. Soc. 134(47), 19350-3, (2012)

Loading...


Red blood cells mediate the onset of thrombosis in the ferric chloride murine model. Barr JD, Chauhan AK, Schaeffer GV, et al. Blood 121(18), 3733-41, (2013)

Loading...


Oxidative stability of soybean oil in oleosomes as affected by pH and iron. Kapchie VN, Yao L, Hauck CC, et al. Food Chem. 141(3), 2286-93, (2013)

Loading...


In vitro and in vivo evaluation of the effect of elevated factor VIII on the thrombogenic process. Golder M, Mewburn J, and Lillicrap D Thromb. Haemost. 109(1), 53-60, (2013)

Loading...


GDF-15 prevents platelet integrin activation and thrombus formation. Rossaint J, Vestweber D, and Zarbock A J. Thromb. Haemost. 11(2), 335-44, (2013)

Loading...


Fluoride adsorption properties of three modified forms of activated alumina in drinking water. Duan Y, Wang C, Li X, et al. J. Water Health 12(4), 715-21, (2014)

Loading...


Thymidine phosphorylase participates in platelet signaling and promotes thrombosis. Li W, Gigante A, Perez-Perez MJ, et al. Circ. Res. 115(12), 997-1006, (2014)

Loading...


RUC-4: a novel αIIbβ3 antagonist for prehospital therapy of myocardial infarction. Li J, Vootukuri S, Shang Y, et al. Arterioscler. Thromb. Vasc. Biol. 34(10), 2321-9, (2014)

Loading...


[Influence of transition metal compounds on superoxide dismutase activity of sulfur reducing Desulfuromonas acetoxidans bacteria]. Vasyliv OM and Hnatush SO Mikrobiol. Z. 75(2), 37-44, (2013)

Loading...


Isolation of pathogen-containing vacuoles. Shevchuk O and Steinert M Methods Mol. Biol. 983, 419-29, (2013)

Loading...


Iron alters red blood cell morphology. Pretorius E and Lipinski B Blood 121(1), 9, (2013)

Loading...


Changes of excitation/emission matrixes of wastewater caused by Fenton- and Fenton-like treatment and their associations with the generation of hydroxyl radicals, oxidation of effluent organic matter and degradation of trace-level organic pollutants. Li W, Nanaboina V, Zhou Q, et al. J. Hazard. Mater. 244-245, 698-708, (2013)

Loading...


Relaxivity control of magnetic nanoclusters for efficient magnetic relaxation switching assay. Cha J, Kwon YS, Yoon TJ, et al. Chem. Commun. (Camb.) 49(5), 457-9, (2013)

Loading...


Iron induces bimodal population development by Escherichia coli. DePas WH, Hufnagel DA, Lee JS, et al. Proc. Natl. Acad. Sci. U. S. A. 110(7), 2629-34, (2013)

Loading...


Iron(III) chloride in oxidative C-C coupling reactions. Sarhan AA and Bolm C Chem. Soc. Rev. 38(9), 2730-44, (2009)

Loading...


FeCl3 catalyzed Prins-type cyclization for the synthesis of highly substituted indenes: application to the total synthesis of (±)-jungianol and epi-jungianol. Dethe DH and Murhade G Org. Lett. 15(3), 429-31, (2013)

Loading...


Efficient generation of ortho-naphthoquinone methides from 1,4-epoxy-1,4-dihydronaphthalenes and their annulation with allyl silanes. Sawama Y, Shishido Y, Yanase T, et al. Angew. Chem. Int. Ed. Engl. 52(5), 1515-9, (2013)

Loading...


Formation of iron containing aggregates at the liquid-air interface. Wieland DC, Degen P, Paulus M, et al. Colloids Surf. B Biointerfaces 109, 74-81, (2013)

Loading...


FeCl3-catalyzed cyclization of α-sulfonamido-allenes with aldehydes--the substituent effect. Cheng J, Tang X, Yu Y, et al. Chem. Commun. (Camb.) 48(99), 12074-6, (2012)

Loading...


Production of hybrid diesel fuel precursors from carbohydrates and petrochemicals using formic acid as a reactive solvent. Zhou X and Rauchfuss TB ChemSusChem 6(2), 383-8, (2013)

Loading...


Synthesis, characterization and potential applications of multifunctional PEO-PPOPEO- magnetic drug delivery system. Dehvari K and Lin KS Curr. Med. Chem. 19(30), 5199-204, (2012)

Loading...


FeCl3-mediated Friedel-Crafts hydroarylation with electrophilic N-acetyl indoles for the synthesis of benzofuroindolines. Beaud R, Guillot R, Kouklovsky C, et al. Angew. Chem. Int. Ed. Engl. 51(50), 12546-50, (2012)

Loading...


Desmolaris, a novel factor XIa anticoagulant from the salivary gland of the vampire bat (Desmodus rotundus) inhibits inflammation and thrombosis in vivo. Ma D, Mizurini DM, Assumpção TC, et al. Blood 122(25), 4094-106, (2013)

Loading...


Source-dependent intracellular distribution of iron in lens epithelial cells cultured under normoxic and hypoxic conditions. Goralska M, Nagar S, Fleisher LN, et al. Invest. Ophthalmol. Vis. Sci. 54(12), 7666-73, (2013)

Loading...


Pretreatment of olive tree biomass with FeCl3 prior enzymatic hydrolysis. López-Linares JC, Romero I, Moya M, et al. Bioresour. Technol. 128, 180-7, (2013)

Loading...


Zinc transferrin stimulates red blood cell formation in the head kidney of common carp (Cyprinus carpio). Chen YH, Fang SW, and Jeng SS Comp. Biochem. Physiol. A. Mol. Integr. Physiol. 166(1), 1-7, (2013)

Loading...


Regulation of HFE expression by poly(ADP-ribose) polymerase-1 (PARP1) through an inverted repeat DNA sequence in the distal promoter. Pelham C, Jimenez T, Rodova M, et al. Biochim. Biophys. Acta 1829(12), 1257-65, (2013)

Loading...


Iron halide-mediated regio- and stereoselective halosulfonylation of terminal alkynes with sulfonylhydrazides: synthesis of (E)-β-chloro and bromo vinylsulfones. Li X, Shi X, Fang M, et al. J. Org. Chem. 78(18), 9499-504, (2013)

Loading...


Acid sphingomyelinase regulates platelet cell membrane scrambling, secretion, and thrombus formation. Münzer P, Borst O, Walker B, et al. Arterioscler. Thromb. Vasc. Biol. 34(1), 61-71, (2014)

Loading...


Deferasirox shows in vitro and in vivo antileukemic effects on murine leukemic cell lines regardless of iron status. Lee DH, Jang PS, Chung NG, et al. Exp. Hematol. 41(6), 539-46, (2013)

Loading...


Characterization of cake layer structure on the microfiltration membrane permeability by iron pre-coagulation. Wang J, Pan S, and Luo D J. Environ. Sci. (China) 25(2), 308-15, (2013)

Loading...


Troubleshooting the rabbit ferric chloride-induced arterial model of thrombosis to assess in vivo efficacy of antithrombotic drugs. Couture L, Richer LP, Mercier M, et al. J. Pharmacol. Toxicol. Methods 67(2), 91-7, (2013)

Loading...


Flocculation optimization of microalga Nannochloropsis oculata. Shen Y, Cui Y, and Yuan W Appl. Biochem. Biotechnol. 169(7), 2049-63, (2013)

Loading...


Elevated ferric, calcium and magnesium ions in the brain induce protein aggregation in brain mitochondria. Alleyne T, Mohan N, and Adogwa A West Indian Med. J. 61(2), 122-7, (2012)

Loading...


Emergency do not consume/do not use concentrations for ferric chloride in drinking water. Willhite CC, Ball GL, and Bhat VS Hum. Exp. Toxicol. 32(3), 260-74, (2013)

Loading...


In vitro anti-oxidative activities and GC-MS analysis of various solvent extracts of Cassia singueana parts. Ibrahim MA, Koorbanally NA, and Islam MS Acta Pol. Pharm. 70(4), 709-19, (2013)

Loading...


Proteasome proteolysis supports stimulated platelet function and thrombosis. Gupta N, Li W, Willard B, et al. Arterioscler. Thromb. Vasc. Biol. 34(1), 160-8, (2014)

Loading...


Intravenous ferric chloride hexahydrate supplementation induced endothelial dysfunction and increased cardiovascular risk among hemodialysis patients. Kuo KL, Hung SC, Lin YP, et al. PLoS ONE 7(12), e50295, (2012)

Loading...


Merck 14,4019

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?