80642 Fluka

Piperidine

purum, ≥96.0% (GC/T)

DOWNLOAD MSDS (PDF)

Synonym: Hexahydropyridine

  • CAS Number 110-89-4

  • Empirical Formula (Hill Notation) C5H11N

  • Molecular Weight 85.15

  •  Beilstein Registry Number 102438

  •  EC Number 203-813-0

  •  MDL number MFCD00005979

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Building Blocks, C5 to C7, Chemical Synthesis, Heterocyclic Building Blocks, Piperidines More...
vapor density   3 (vs air)
vapor pressure   23 mmHg ( 20 °C)
grade   purum
assay   ≥96.0% (GC/T)
impurities   ≤0.5% water
refractive index   n20/D 1.452(lit.)
  n20/D 1.453
bp   106 °C(lit.)
mp   −13 °C(lit.)
density   0.862 g/mL at 20 °C(lit.)

Price and Availability

Customers Also Viewed

Piperidine

≥99.5%, purified by redistillation

Piperidine

biotech. grade, ≥99.5%

Piperidine

puriss. p.a., ≥99.0% (GC/T)

Piperidine

ReagentPlus®, 99%

Piperidine

puriss. plus, ≥99.5% (GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?