80645 Fluka

Piperidine solution

for peptide synthesis, 20% in DMF

DOWNLOAD MSDS (PDF)
  • CAS Number 110-89-4

  • Empirical Formula (Hill Notation) C5H11N

  • Molecular Weight 85.15

  •  Beilstein Registry Number 1421082

  •  MDL number MFCD00005979

  •  PubChem Substance ID 24887574

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Chemical Biology, Chemical Synthesis, Peptide Chemistry, Solvents and Mixtures for Peptide Synthesis
grade   for peptide synthesis
concentration   20% in DMF
impurities   ≤0.1% water
refractive index   n20/D 1.434
density   0.930 g/mL at 20 °C

Description

Other Notes

Sales restrictions may apply

Packaging

2 L in glass bottle

500 mL in glass bottle

Price and Availability

Customers Also Viewed

Piperidine

≥99.5%, purified by redistillation

Piperidine

biotech. grade, ≥99.5%

Piperidine

puriss. p.a., ≥99.0% (GC/T)

Piperidine

ReagentPlus®, 99%

Piperidine

puriss. plus, ≥99.5% (GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?