Synonym:
4,4-
CAS Number 40064-34-4
Empirical Formula (Hill Notation) C5H11NO2 · HCl
Molecular Weight 153.61
Beilstein Registry Number 3916558
EC Number 254-779-9
MDL number MFCD00012776
PubChem Substance ID 24887582
Popular Documents: FTNMR (PDF)
| Related Categories | Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Piperidones |
| grade | purum |
| assay | ≥98.0% (AT) |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?