Synonym: Disodium hydrogen phosphate solution, Sodium phosphate dibasic solution, di-Sodium hydrogen phosphate solution
CAS Number 7558-79-4
Linear Formula Na2HPO4
Molecular Weight 141.96
EC Number 231-791-2
MDL number MFCD00003496
PubChem Substance ID 24887682
| grade | for ion chromatography |
| quality | certified by BAM, according to ISO 17025 |
| concentration | 1000 mg/kg PO43- |
Exact concentration, expiry date, traceability and detailed information on certification can be found on the certificate and certification report, included in each package. Content and expiry date on label.
prepared with high purity Na2HPO4 and high purity water (18 MOhm, filtered 0.22 μm)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?