Synonym: PEG
CAS Number 25322-68-3
Linear Formula H(OCH2CH2)nOH
MDL number MFCD00081839
PubChem Substance ID 24887690
Popular Documents: FTIR (PDF)
| Related Categories | Essential Chemicals, Inorganic Salts, Solutions and Reagents, Materials Science, PEG and Oligo Ethylene Glycol, Poly(ethylene glycol), |
| vapor density | >1 (vs air) |
| vapor pressure | <0.01 mmHg ( 20 °C) |
| autoignition temp. | 581 °F |
| mol wt | average Mw 1,500 |
| mp | 45-50 °C |
| transition temp | Tm 45-50 °C |
1 kg in poly bottle
5 kg in glass bottle
Molecular weight range: 1,400-1,600
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?