EMAIL THIS PAGE TO A FRIEND

81440 Fluka

Polyvinylpyrrolidone

K 90, powder, average Mw 360,000

Synonym: PVP, Polyvidone, Povidone

Purchase

Description

Other Notes

Polyvinylpyrrolidone is a component of Denhardt′s Solution and is included at a concentration of 1% (w/v) in the standard 50X stock solution.

Packaging

1 kg in poly bottle

250 g in poly bottle

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.




Custom Polymer Synthesis

Customers Also Viewed

Polyvinylpyrrolidone

average mol wt 360,000

Polyvinylpyrrolidone

for molecular biology, nucleic acid hybridization tested, mol wt 360,000

Polyvinylpyrrolidone

K 30, average Mw 40,000

Polyvinylpyrrolidone

average Mw ~1,300,000 by LS

Polyvinylpyrrolidone solution

K 60, 45% in H2O, Mr ~160000

Safety & Documentation

Safety Information

WGK Germany 
1
RTECS 
TR8370000

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Th2 cytokines associated with chronic rhinosinusitis with polyps down-regulate the antimicrobial immune function of human sinonasal epithelial cells. Murugappan Ramanathan et al Am. J. Rhinol. 22, 115-21, (2010)

Loading...


Classification of Cryptosporidium species from patients with sporadic cryptosporidiosis by use of sequence-based multilocus analysis following mutation scanning. Jex AR J. Clin. Microbiol. 46(7), 2252-62, (2008)

Loading...


Solid dispersion of quercetin in cellulose derivative matrices influences both solubility and stability. Li B, Konecke S, Harich K, et al. Carbohydr. Polym. 92(2), 2033-40, (2013)

Loading...


Regulation of sodium and calcium in Daphnia magna exposed to silver nanoparticles. Zhao CM and Wang WX Environ. Toxicol. Chem. 32(4), 913-9, (2013)

Loading...


The selective serotonin reuptake inhibitor sertraline enhances counterregulatory responses to hypoglycemia. Sanders, NM Am. J. Physiol. Endocrinol. Metab. 294(5), E853-60, (2008)

Loading...


[Gordon's polyvinylpyrrolidone test]. Aebersold J Helv. Med. Acta 32(3), 227-46, (1965)

Loading...


Removal of phenolic compounds from aqueous phase by adsorption onto polymer supported iron nanoparticles. Sen BK, Deshmukh DK, Deb MK, et al. Bull. Environ. Contam. Toxicol. 93(5), 549-54, (2014)

Loading...


A systematic evaluation of agglomeration of Ag and TiO2 nanoparticles under freshwater relevant conditions. Topuz E, Sigg L, and Talinli I Environ. Pollut. 193, 37-44, (2014)

Loading...


[Antimicrobial activity of stable silver nanoparticles of a certain size]. Mukha IuP, Eremenko AM, Smirnova NP, et al. Prikl. Biokhim. Mikrobiol. 49(2), 215-23, (2013)

Loading...


Role of tentacles and protein loading on pore accessibility and mass transfer in cation exchange materials for proteins. Thomas H, Coquebert de Neuville B, Storti G, et al. J. Chromatogr. A 1285, 48-56, (2013)

Loading...


Electromechanical resonator based on electrostatically actuated graphene-doped PVP nanofibers. Fardindoost S, Mohammadi S, Iraji zad A, et al. Nanotechnology 24(13), 135201, (2013)

Loading...


[Jumper's knee. Surgery and arthroscopic treatment with scraping and povidone collagen in high-performance athletes]. Cuéllar EC and Mina NZ Acta Ortop. Mex. 21(5), 234-8, (2007)

Loading...


A physiological replacement for polyvinylpyrrolidone (PVP) in assisted reproductive technology. Barak Y, Menezo Y, Veiga A, et al. Hum. Fertil. (Camb.) 4(2), 99-103, (2001)

Loading...


Drug "supersaturation" states induced by polymeric micelles and liposomes: a mechanistic investigation into permeability enhancements. di Cagno M and Luppi B Eur. J. Pharm. Sci. 48(4-5), 775-80, (2013)

Loading...


Modifying excitation light dose of novel photosensitizer PVP-Hypericin for photodynamic diagnosis and therapy. Penjweini R, Loew HG, Eisenbauer M, et al. J. Photochem. Photobiol. B, Biol. 120, 120-9, (2013)

Loading...


A method for nanofluidic device prototyping using elastomeric collapse. Seung-min Park et al Proc. Natl. Acad. Sci. U. S. A. 106, 15549-54, (2009)

Loading...


Ultralow-limit gas detection in nano-dumbbell polymer sensor via electrospinning. Xue M, Li F, Wang Y, et al. Nanoscale 5(5), 1803-5, (2013)

Loading...


Effect of drug-polymer interactions on the aqueous solubility of milled solid dispersions. Al-Obaidi H, Lawrence MJ, Shah S, et al. Int. J. Pharm. 446(1-2), 100-5, (2013)

Loading...


NIR imaging spectroscopy for quantification of constituents in polymers thin films loaded with paracetamol. Alexandrino GL and Poppi RJ Anal. Chim. Acta 765, 37-44, (2013)

Loading...


Preparation of poly(phthalazinone-ether-sulfone) sponge-like ultrafiltration membrane. Qin P, Hong X, Karim MN, et al. Langmuir 29(12), 4167-75, (2013)

Loading...


Genotoxicity of polyvinylpyrrolidone-coated silver nanoparticles in BEAS 2B cells. Nymark P, Catalán J, Suhonen S, et al. Toxicology 313(1), 38-48, (2013)

Loading...


Comparing the effects of nanosilver size and coating variations on bioavailability, internalization, and elimination, using Lumbriculus variegatus. Coleman JG, Kennedy AJ, Bednar AJ, et al. Environ. Toxicol. Chem. 32(9), 2069-77, (2013)

Loading...


Polymer crowders and protein crowders act similarly on protein folding stability. Zhou HX FEBS Lett. 587(5), 394-7, (2013)

Loading...


Silver nanoparticle toxicity to Daphnia magna is a function of dissolved silver concentration. Newton KM, Puppala HL, Kitchens CL, et al. Environ. Toxicol. Chem. 32(10), 2356-64, (2013)

Loading...


Comprehensive analysis of alterations in the miRNome in response to photodynamic treatment. Bach D, Fuereder J, Karbiener M, et al. J. Photochem. Photobiol. B, Biol. 120, 74-81, (2013)

Loading...


Large-scale and highly efficient synthesis of micro- and nano-fibers with controlled fiber morphology by centrifugal jet spinning for tissue regeneration. Ren L, Pandit V, Elkin J, et al. Nanoscale 5(6), 2337-45, (2013)

Loading...


Synthesis and chirality control of bulk crystals and nanocrystals: from a right-handed nonpolar chain to a left-handed polar chain. Yin YY, Zhao YL, Ma JG, et al. Inorg. Chem. 52(7), 3738-43, (2013)

Loading...


An investigation into the effect of spray drying temperature and atomizing conditions on miscibility, physical stability, and performance of naproxen-PVP K 25 solid dispersions. Paudel A, Loyson Y, and Van den Mooter G J. Pharm. Sci. 102(4), 1249-67, (2013)

Loading...


Detecting phase separation of freeze-dried binary amorphous systems using pair-wise distribution function and multivariate data analysis. Chieng N, Trnka H, Boetker J, et al. Int. J. Pharm. 454(1), 167-73, (2013)

Loading...


Dissolution testing of amorphous solid dispersions. Kogermann K, Penkina A, Predbannikova K, et al. Int. J. Pharm. 444(1-2), 40-6, (2013)

Loading...


Folic acid-conjugated graphene oxide for cancer targeted chemo-photothermal therapy. Qin XC, Guo ZY, Liu ZM, et al. J. Photochem. Photobiol. B, Biol. 120, 156-62, (2013)

Loading...


Anaphylaxis to polyvinylpyrrolidone after vaginal application of povidone-iodine. Adachi A, Fukunaga A, Hayashi K, et al. Contact Dermatitis 48(3), 133-6, (2003)

Loading...


Mechanochemical activation of vincamine mediated by linear polymers: assessment of some "critical" steps. Hasa D, Perissutti B, Grassi M, et al. Eur. J. Pharm. Sci. 50(1), 56-68, (2013)

Loading...


II. Technological approaches to improve the dissolution behavior of nateglinide, a lipophilic insoluble drug: co-milling. Maggi L, Bruni G, Maietta M, et al. Int. J. Pharm. 454(1), 568-72, (2013)

Loading...


Water-soluble and biocompatible MnO@PVP nanoparticles for MR imaging in vitro and in vivo. Hu X, Ji Y, Wang M, et al. J. Biomed. Nanotechnol. 9(6), 976-84, (2013)

Loading...


Removal of polyvinylpyrrolidone from wastewater using different methods. Julinová M, Kupec J, Houser J, et al. Water Environ. Res. 84(12), 2123-32, (2012)

Loading...


Polyvinylpyrrolidone-sodium hyaluronate gel (Gelclair): a bioadherent oral gel for the treatment of oral mucositis and other painful oral lesions. Buchsel PC Expert Opin. Drug Metab. Toxicol. 4(11), 1449-54, (2008)

Loading...


Solid dispersions of the penta-ethyl ester prodrug of diethylenetriaminepentaacetic acid (DTPA): formulation design and optimization studies. Yang YT, Di Pasqua AJ, Zhang Y, et al. Pharm. Dev. Technol. 19(7), 806-12, (2014)

Loading...


Investigating oxidative stress and inflammatory responses elicited by silver nanoparticles using high-throughput reporter genes in HepG2 cells: effect of size, surface coating, and intracellular uptake. Prasad RY, McGee JK, Killius MG, et al. Toxicol. In Vitro 27(6), 2013-21, (2013)

Loading...


Biochemical and hematologic effects of polyvinylpyrrolidone-wrapped fullerene C60 after oral administration. Yamashita K, Yoshioka Y, Pan H, et al. Pharmazie 68(1), 54-7, (2013)

Loading...


Curcumin amorphous solid dispersions: the influence of intra and intermolecular bonding on physical stability. Wegiel LA, Zhao Y, Mauer LJ, et al. Pharm. Dev. Technol. 19(8), 976-86, (2014)

Loading...


Chemical and microbiologic characteristics and toxicity of povidone-iodine solutions. Zamora JL Am. J. Surg. 151(3), 400-6, (1986)

Loading...


Phosphate-enhanced cytotoxicity of zinc oxide nanoparticles and agglomerates. Everett WN, Chern C, Sun D, et al. Toxicol. Lett. 225(1), 177-84, (2014)

Loading...


Composite nanofibers doped with a phosphorescent Re(I) complex having restricted conjugation plane in its diamine ligand: preparation, characterization, photophysical property and oxygen-sensing performance. Lin C, Shaoyan W, Qi W, et al. Spectrochim. Acta. A. Mol. Biomol. Spectrosc. 114, 330-8, (2013)

Loading...


Solution growth of spherulitic rod and platelet calcium phosphate assemblies through polymer-assisted mesoscopic transformations. Kosma VA and Beltsios KG Mater. Sci. Eng. C. Mater. Biol. Appl. 33(4), 2175-91, (2013)

Loading...


A novel surface-attached carvedilol solid dispersion with enhanced solubility and dissolution. Lee SN, Poudel BK, Tran TH, et al. Arch. Pharm. Res. 36(1), 79-85, (2013)

Loading...


Direct and indirect method of plant regeneration from root explants of Caesalpinia bonduc (L.) Roxb.--a threatened medicinal plant of Western Ghats. Kumar SR, Krishna V, Venkatesh, et al. Indian J. Exp. Biol. 50(12), 910-7, (2012)

Loading...


Powder flow in an automated uniaxial tester and an annular shear cell: a study of pharmaceutical excipients and analytical data comparison. Kuentz M and Schirg P Drug Dev. Ind. Pharm. 39(9), 1476-83, (2013)

Loading...


Novel taste-masked orally disintegrating tablets for a highly soluble drug with an extremely bitter taste: design rationale and evaluation. Tan Q, Zhang L, Liu G, et al. Drug Dev. Ind. Pharm. 39(9), 1364-71, (2013)

Loading...


A non-destructive genotyping system from a single seed for marker-assisted selection in watermelon. Meru G, McDowell D, Waters V, et al. Genet. Mol. Res. 12(1), 702-9, (2013)

Loading...


Preparation, characterization and tableting of cilnidipine solid dispersions. Hu L, Song W, Niu F, et al. Pak. J. Pharm. Sci. 26(3), 629-36, (2013)

Loading...


Formulation and evaluation of clozapine orally disintegrating tablets prepared by direct compression. Olmez SS, Vural I, Sahin S, et al. Pharmazie 68(2), 110-6, (2013)

Loading...


Polyvinylpyrrolidone (PVP), its diagnostic, therapeutic and technical application and consequences thereof. Wessel W, Schoog M, and Winkler E Arzneimittelforschung 21(10), 1468-82, (1971)

Loading...


Povidone-iodine solutions in traumatic wound preparation. Oberg MS Am. J. Emerg. Med. 5(6), 553-5, (1987)

Loading...


Transport and accumulation of PVP-Hypericin in cancer and normal cells characterized by image correlation spectroscopy techniques. Penjweini R, Smisdom N, Deville S, et al. Biochim. Biophys. Acta 1843(5), 855-65, (2014)

Loading...


Fast disintegrating tablets of nisoldipine for intra-oral administration. El Maghraby GM and Elsergany RN Pharm. Dev. Technol. 19(6), 641-50, (2014)

Loading...


The influence of sodium salts (iodide, chloride and sulfate) on the formation efficiency of sulfamerazine nanocrystals. Lou H, Liu M, Qu W, et al. Pharm. Dev. Technol. 19(5), 548-55, (2014)

Loading...


Merck 14,7697

Aldrich MSDS 1, 1530:C / Corp MSDS 1 (2), 2892:D / FT-IR 2 (3), 4701:D / FT-IR 1 (2), 1202:C / FT-IR 1 (2), 1202:D / FT-IR 2 (3), 4702:B / FT-IR 1 (2), 1202:A / FT-IR 2 (3), 4702:A / IR-Spectra (3), 1592:F / IR-Spectra (2), 1354:G / IR-Spectra (2), 1354:F / IR-Spectra (3), 1592:D / IR-Spectra (3), 1592:E / RegBook 1 (2), 3171:F / Sax 6, 2265 / Sigma FT-IR 1 (1), 702:A / Sigma FT-IR 1 (1), 701:D / Structure Index 1, 504:B:1

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?