• USA Home
  • 82313 - (R)-(+)-Propylene oxide

EMAIL THIS PAGE TO A FRIEND

82313 Fluka

(R)-(+)-Propylene oxide

puriss., ≥99.0% (sum of enantiomers, GC)

Synonym: (R)-(+)-Methyloxirane, (R)-(+)-1,2-Epoxypropane

  • CAS Number 15448-47-2

  • Empirical Formula (Hill Notation) C3H6O

  • Molecular Weight 58.08

  •  Beilstein Registry Number 79764

  •  MDL number MFCD00066211

  • Popular Documents:  FTIR (PDF)  

Purchase

Properties

grade   puriss.
assay   ≥99.0% (sum of enantiomers, GC)
optical activity   [α]20/D +14±1°, neat
refractive index   n20/D 1.366(lit.)
  n20/D 1.366
bp   33-34 °C(lit.)
density   0.829 g/mL at 20 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

Chiral building block for introducing the (R)-2-hydroxypropyl group by epoxide opening with carbon nucleophiles 1,2,3

Price and Availability

Customers Also Viewed

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Hazard Codes (Europe) 
Risk Statements (Europe) 
Safety Statements (Europe) 
45
RIDADR 
UN 1280 3 / PGI
WGK Germany 
3
RTECS 
UJ2650000
Flash Point(F) 
-34.6 °F
Flash Point(C) 
-37 °C

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

1. J.D. White et al. J. Am. Chem. Soc. 108, 5352, (1986)

2. R. Whitby, P. Kocienski J. Chem. Soc. Chem. Commun., 906, (1987)

3. F.M.C. Leemhuis et al. J. Org. Chem. 58, 7170, (1993)

Merck 13,7948

Beil. 17,V,1,16

FT-IR 2 (1), 358:A

Related Products

replaced by

Product #

Description

Add to Cart

540048 (R)-(+)-Propylene oxide, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?