• USA Home
  • 82562 - Buffer tablets pH 9.2

EMAIL THIS PAGE TO A FRIEND
82562 Fluka

Buffer tablets pH 9.2

for 100 mL solution

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Buffer Tablets & Powders, Titration, pH Buffers for Titration More...
impurities   disodium tetraborate

Description

General description

1 pack contains 50 tablets

Visit our Titration Center to learn more.

Legal Information

FLUKA is a registered trademark of Honeywell Specialty Chemicals Seelze GmbH

Reconstitution

one tablet dissolved in distilled water and made up to 100 mL produces a solution of pH 9.2 at 20°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?