83374 Fluka

Mercury(II) thiocyanate

purum p.a., ≥97.0% (precipitation titration)

DOWNLOAD MSDS (PDF)

Synonym: Mercuric isothiocyanate, Mercuric sulfocyanide, Mercuric thiocyanate, Mercury di(thiocyanate), Thiocyanic acid, mercury(II) salt

  • CAS Number 592-85-8

  • Linear Formula Hg(SCN)2

  • Molecular Weight 316.75

  •  Beilstein Registry Number 3561344

  •  EC Number 209-773-0

  •  MDL number MFCD00011039

  •  PubChem Substance ID 24887939

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Mercury More...
grade   purum p.a.
assay   ≥97.0% (precipitation titration)
mp   165 °C (dec.)(lit.)
density   3.71 g/mL at 25 °C(lit.)
cation traces   Ca: ≤50 mg/kg
  Cd: ≤50 mg/kg
  Co: ≤50 mg/kg
  Cu: ≤50 mg/kg
  Fe: ≤50 mg/kg
  K: ≤1000 mg/kg
  Na: ≤100 mg/kg
  Ni: ≤50 mg/kg
  Pb: ≤50 mg/kg
  Zn: ≤50 mg/kg

Description

Application

Catalyst for addition of HSCN to alkynes.

Price and Availability

Customers Also Viewed

Iron(III) nitrate nonahydrate

ACS reagent, ≥98%

Mercury(II) thiocyanate

99.99% trace metals basis

Mercury(II) thiocyanate

96.5-103.5% (titration)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?