83909 Fluka

Rifamycin SV sodium salt

≥98.0% (HPLC), ≥900 U/mg dried material

DOWNLOAD MSDS (PDF)
  • CAS Number 14897-39-3

  • Empirical Formula (Hill Notation) C37H46NNaO12

  • Molecular Weight 719.75

  •  EC Number 238-965-7

Description

Other Notes

A reducing substitute, such as ascorbic acid, should be added to aqueous solutions of rifamycin SV to prevent its transformation to rifamycin. Inhibitor of bacterial DNA-dependent RNA-polymerase1

Unit Definition

1 U corresponds to 1 μg of the Ph Eur rifamycin reference standard

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Customers Also Viewed

Rifampicin

≥97% (HPLC), powder

Rifampicin

plant cell culture tested, BioReagent, ≥97% (HPLC), crystalline

Rifampicin

powder, γ-irradiated

Rifamycin SV sodium salt

potency: ≥900 units (dry basis)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?