84260 Fluka

Benzyl salicylate

purum, ≥99.0% (GC)

DOWNLOAD MSDS (PDF)

Synonym: Benzyl 2-hydroxybenzoate

  • CAS Number 118-58-1

  • Linear Formula 2-(HO)C6H4CO2CH2C6H5

  • Molecular Weight 228.24

  •  Beilstein Registry Number 2115365

  •  EC Number 204-262-9

  •  MDL number MFCD00020034

  •  PubChem Substance ID 24888191

Properties

Related Categories Building Blocks, C12 to C63, Carbonyl Compounds, Chemical Synthesis, Esters,
grade   purum
assay   ≥99.0% (GC)
refractive index   n20/D 1.581(lit.)
  n20/D 1.581
bp   168-170 °C/5 mmHg(lit.)
acid value   ≤5
transition temp   solidification point 24 °C(lit.)
density   1.176 g/mL at 25 °C(lit.)

Price and Availability

Customers Also Viewed

Benzyl salicylate

≥98%

Methyl salicylate

ReagentPlus®, ≥99% (GC)

Salicylic acid

BioXtra, ≥99.0%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?