Synonym: 2-Propenoic acid,2-methyl homopolymer, 2-methyl-2-propenoic acid homopolymer, Microparticles based on PMAA, Microparticles based on poly(methacrylic acid), Polymethacrylate
MDL number MFCD00198073
| Related Categories | Biochemicals and Reagents, Materials Science, Microparticles, Microparticles based on polymethacrylate, Particle Size Standards, |
| form | suspension |
| concentration | 10% (solids) |
| particle size | 40 μm std dev <0.8 μm, coeff var <2% |
| storage temp. | 2-8°C |
The exact values for particle size and standard deviation are determined for each lot and may be found on the certificate of analysis. These values are determined with an accuracy of 0.01 μm (Coulter Multisizer). Polymethacrylate based particles have a narrow size distribution and spherical shape. They are stable at temperatures of up to 200 °C. Density is 1.22 g/cm3. The surface is negatively charged. Good biocompatibility and reduced nonspecific protein binding are characteristic.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?