Synonym:
1-
CAS Number 57-11-4
Linear Formula CH3(CH2)16COOH
Molecular Weight 284.48
Beilstein Registry Number 608585
EC Number 200-313-4
MDL number MFCD00002752
Popular Documents: FTNMR (PDF)
| Related Categories | Allium cepa (Onion), Aloe Vera, Biochemicals and Reagents, Building Blocks, C13 to C42+, |
| vapor pressure | 1 mmHg ( 173.7 °C) |
| grade | puriss. |
| assay | ≥98.5% (GC) |
| ign. residue | ≤0.05% |
| bp | 361 °C(lit.) |
| mp | 67-72 °C(lit.) |
| 69-71 °C |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?