• USA Home
  • 85975 - N-Succinyl-Ala-Ala-Ala-p-nitroanilide

EMAIL THIS PAGE TO A FRIEND

85975 Fluka

N-Succinyl-Ala-Ala-Ala-p-nitroanilide

≥99.0% (enzymatic)

Synonym: N-Succinyl-L-alanyl-L-alanyl-L-alanine 4-nitroanilide, N-Succinyl-tri-L-alanine 4-nitroanilide

  • CAS Number 52299-14-6

  • Empirical Formula (Hill Notation) C19H25N5O8

  • Molecular Weight 451.43

  •  Beilstein Registry Number 2928351

  •  EC Number 257-823-5

  •  MDL number MFCD00036774

Purchase

Properties

assay   ≥99.0% (enzymatic)
storage temp.   2-8°C

Description

Other Notes

Chromogenic substrate for the assay of pancreatic elastase1,2

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

1. R. Geiger, H.U. Bergmeyer, ed. Methods of Enzymatic Analysis 3rd ed., Weinheim 5, 170, (1984)

2. The significance of the N-acyl residue of L-alanyl-L-alanyl-L-alanine p-nitroanilide for cleavage by pancreatic elastase. E. Kasafirek et al. FEBS Lett. 40, 353, (1974)

Loading...


Related Products

replaced by

Product #

Description

Add to Cart

S4760 N-Succinyl-Ala-Ala-Ala-p-nitroanilide, elastase substrate

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?