EMAIL THIS PAGE TO A FRIEND

86410 Fluka

Terephthalaldehyde

purum, ≥98.0% (HPLC)

Synonym: Benzene-1,4-dicarboxaldehyde, Terephthaldicarboxaldehyde

Purchase

Properties

Related Categories Aldehydes, Building Blocks, C8, Carbonyl Compounds, Chemical Synthesis,
grade   purum
assay   ≥98.0% (HPLC)
impurities   ≤1% water
bp   245-248 °C(lit.)
mp   114-116 °C(lit.)
  114-116 °C

Description

Other Notes

This product has been replaced by T2207-ALDRICH | Terephthalaldehyde ReagentPlus, 99%

Price and Availability


Search for Answers, Not Just Materials

Safety & Documentation

Safety Information

WGK Germany 
1
RTECS 
WZ0430000
Flash Point(F) 
168.8 °F
Flash Point(C) 
76 °C

Protocols & Articles

Peer-Reviewed Papers

References

Set your institution to view full text papers.

Beil. 7,IV,2140

FT-IR 2 (2), 2533:A / FT-IR 1 (2), 113:A / FT-NMR 1 (2), 945:B / IR-Spectra (2), 801:G / IR-Spectra (3), 916:A / NMR-Reference 2 (2), 111:C / RegBook 1 (2), 1719:G / Sax 6, 2503 / Structure Index 1, 273:B:2 / Vapor Phase 3, 1293:A

Related Products

replaced by

Product #

Description

Add to Cart

T2207 Terephthalaldehyde, ReagentPlus®, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?