86606 Fluka

Tetraethylammonium bromide

for electrochemical analysis, ≥99.0%

DOWNLOAD MSDS (PDF)

Synonym: TEA bromide

  • CAS Number 71-91-0

  • Linear Formula (C2H5)4N(Br)

  • Molecular Weight 210.16

  •  Beilstein Registry Number 3563430

  •  EC Number 200-769-4

  •  MDL number MFCD00011825

  •  PubChem Substance ID 24888701

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Ammonium Salts, Analytical Reagents, Analytical/Chromatography, Chemical Synthesis, Electrochemistry,
grade   for electrochemical analysis
assay   ≥99.0% (AT)
  ≥99.0%
mp   285 °C (dec.)(lit.)
solubility   acetonitrile: soluble0.1 g/mL, clear, colorless (warm)

Description

General description

Visit our Sensor Applications portal to learn more.

Price and Availability

Customers Also Viewed

Tetraethylammonium bromide

ReagentPlus®, 99%

Tetraethylammonium bromide

reagent grade, 98%

Tetraethylammonium bromide

for ion pair chromatography, ≥99.0%

Tetraethylammonium bromide

puriss., ≥99.0% (AT)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?