86608 Fluka

Tetraethylammonium bromide

for ion pair chromatography, ≥99.0%

DOWNLOAD MSDS (PDF)

Synonym: TEA bromide

  • CAS Number 71-91-0

  • Linear Formula (C2H5)4N(Br)

  • Molecular Weight 210.16

  •  Beilstein Registry Number 3563430

  •  EC Number 200-769-4

  •  MDL number MFCD00011825

  •  PubChem Substance ID 24888703

  • Popular Documents:  FTNMR (PDF)  

Properties

Related Categories Acidic Ion Pair Reagents (Anionic), Analytical Reagents, Analytical/Chromatography, Buffers & Reagents, Chromatography Reagents & Solvents,
grade   for ion pair chromatography
assay   ≥99.0% (AT)
  ≥99.0%
mp   285 °C (dec.)(lit.)
λ   10 % in H2O
UV absorption   λ: 240 nm Amax: 0.04
  λ: 250 nm Amax: 0.03
  λ: 260 nm Amax: 0.02
  λ: 500 nm Amax: 0.02
suitability   corresponds to standard for filter test

Price and Availability

Customers Also Viewed

Tetraethylammonium bromide

ReagentPlus®, 99%

Tetraethylammonium bromide

reagent grade, 98%

Tetraethylammonium bromide

for electrochemical analysis, ≥99.0%

Triethylammonium bicarbonate buffer

volatile buffer, for HPLC, 1 M

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?