86627 Fluka

Tetraethylammonium hexafluorophosphate

purum, ≥98.0% (T)

DOWNLOAD MSDS (PDF)
  • CAS Number 429-07-2

  • Linear Formula (C2H5)4N(PF6)

  • Molecular Weight 275.22

  •  Beilstein Registry Number 3748868

  •  EC Number 207-056-7

  •  MDL number MFCD00043179

  •  PubChem Substance ID 24888712

Properties

Related Categories Ammonium Salts, Chemical Synthesis, Phase Transfer Catalysts, Synthetic Reagents
grade   purum
assay   ≥98.0% (T)
mp   ≥300 °C(lit.)

Price and Availability

Customers Also Viewed

Tetraethylammonium hexafluorophosphate

for electrochemical analysis, ≥99.0%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?