CAS Number 210230-40-3
Linear Formula (CH3CH2CH2CH2)4N[N(SO2CF3)2
Molecular Weight 522.61
Beilstein Registry Number 8247973
MDL number MFCD01863113
PubChem Substance ID 24888774
| Related Categories | Ammonium, Analytical Reagents, Analytical/Chromatography, Chemical Synthesis, Electrochemistry, |
| grade | for electronic purposes |
| assay | ≥99.0% (T) |
| ≥99.0% | |
| mp | 90-95 °C |
Visit our Sensor Applications portal to learn more.
Novel type of electrolyte, used e.g. in electrical double layer capacitors1
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?